Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-403
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD34 molecule
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:4H11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P28906
- Antigen synonyms:CD34
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The 4H11 monoclonal antibody specifically reacts with the Class III epitope of CD34 (mucosialin), a transmembrane phosphoglycoprotein. It is expressed on most hematopoietic progenitors, pluripotent stem cells and tumor cells.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human