Anti-SAA4 Rabbit Polyclonal Antibody
Catalog # PRSI58-909
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Serum amyloid A-4 protein
- Antigen symbol:SAA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:97 - 124 aa
- Form:Liquid
- Gene ID:6291
- Modification:None
- Molecular weight:15 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SAA4 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 97-124 amino acids from the C-terminal region of human SAA4
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: SAA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 97-124
Host: Rabbit
Isotype: Ig
Reactivity: Human