Anti-USP1 Rabbit Polyclonal Antibody
PRSI58-780
- Antibody type:Primary
- Antigen name:Ubiquitin carboxyl-terminal hydrolase 1
- Antigen symbol:USP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:662 - 690 aa
- Form:Liquid
- Gene ID:7398
- Modification:None
- Molecular weight:88 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,36 mg/ml
- Shipping temperature:4 °C
- Immunogen:This USP1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 662-690 amino acids from the C-terminal region of human USP1
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: USP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 662-690
Host: Rabbit
Isotype: Ig
Reactivity: Human