Anti-NR2F6 Rabbit Polyclonal Antibody
Catalog # PRSI58-766
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Nuclear receptor subfamily 2 group F member 6
- Antigen symbol:NR2F6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:25 - 53 aa
- Form:Liquid
- Gene ID:2063
- Modification:None
- Molecular weight:43 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NR2F6 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 25-53 amino acids from the N-terminal region of human NR2F6
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: NR2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 25-53
Host: Rabbit
Isotype: Ig
Reactivity: Human