Anti-TTC8 Rabbit Polyclonal Antibody
PRSI58-745
New Product
- Antibody type:Primary
- Antigen name:Tetratricopeptide repeat protein 8
- Antigen symbol:TTC8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:18 - 46 aa
- Form:Liquid
- Gene ID:123016
- Modification:None
- Molecular weight:62 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This TTC8 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 18-46 amino acids from the N-terminal region of human TTC8
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: TTC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 18-46
Host: Rabbit
Isotype: Ig
Reactivity: Human