Anti-EPOR Rabbit Polyclonal Antibody
Catalog # PRSI58-645
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Erythropoietin receptor
- Antigen-Symbol:EPOR
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Durchflusszytometrie:Yes
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Reaktivität:Human
- Western blot:Yes
- Epitop:329 - 357 aa
- Form:Liquid
- Gen-ID:2057
- Modification:None
- Molecular weight:55 kDa
- Lagertemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Konzentration:0,3 mg/ml
- Transporttemperatur:4 °C
- Immunogen:This EPOR antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 329-357 amino acids from the Central region of human EPOR
- Reinigung:This antibody is purified through a protein A column, followed by peptide affinity purification
- VE:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50. For IF starting dilution is: 1:10~50.
Type: Primary
Antigen: EPOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 329-357
Host: Rabbit
Isotype: Ig
Reactivity: Human