Anti-CCDC140 Rabbit Polyclonal Antibody
PRSI58-577
- Antibody type:Primary
- Antigen name:Coiled-coil domain-containing protein 140
- Antigen symbol:CCDC140
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:31 - 60 aa
- Form:Liquid
- Gene ID:151278
- Modification:None
- Molecular weight:18 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CCDC140 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 31-60 amino acids from the N-terminal region of human CCDC140
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: CCDC140
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 31-60
Host: Rabbit
Isotype: Ig
Reactivity: Human