Anti-ARNT2 Rabbit Polyclonal Antibody
Catalog # PRSI58-559
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Aryl hydrocarbon receptor nuclear translocator 2
- Antigen symbol:ARNT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:249 - 278 aa
- Form:Liquid
- Gene ID:9915
- Modification:None
- Molecular weight:79 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ARNT2 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 249-278 amino acids from the Central region of human ARNT2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:10~50.
Type: Primary
Antigen: ARNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 249-278
Host: Rabbit
Isotype: Ig
Reactivity: Human