Anti-GPRC5B Rabbit Polyclonal Antibody
PRSI58-491
New Product
- Antibody type:Primary
- Antigen name:G-protein coupled receptor family C group 5 member B
- Antigen symbol:GPRC5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:293 - 321 aa
- Form:Liquid
- Gene ID:51704
- Modification:None
- Molecular weight:45 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GPRC5B antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 293-321 amino acids from the C-terminal region of human GPRC5B
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GPRC5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 293-321
Host: Rabbit
Isotype: Ig
Reactivity: Human