Anti-NAA20 Rabbit Polyclonal Antibody
PRSI58-467
- Antikörper-Typ:Primär
- Antigen-Name:N-alpha-acetyltransferase 20
- Antigen-Symbol:NAA20
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Epitop:127 - 155 aa
- Form:Liquid
- Gen-ID:51126
- Modification:None
- Molecular weight:20 kDa
- Lagertemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Konzentration:0,5 mg/ml
- Transporttemperatur:4 °C
- Immunogen:This NAT5 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 127-155 amino acids from the C-terminal region of human NAT5
- Reinigung:This antibody is purified through a protein A column, followed by peptide affinity purification
- VE:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: NAA20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 127-155
Host: Rabbit
Isotype: Ig
Reactivity: Human