Anti-Hoxb8 Rabbit Polyclonal Antibody
Codice catalogo PRSI58-242
Nome del fornitore: ProSci Inc.
Specifiche
- Antibody type:Primary
- Antigen name:Homeobox protein Hox-B8
- Antigen symbol:Hoxb8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:214 - 243 aa
- Form:Liquid
- Gene ID:15416
- Modification:None
- Molecular weight:28 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Hoxb8 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 214-243 amino acids from the C-terminal region of mouse Hoxb8
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifiche
Informazioni su questo articolo
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: Hoxb8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 214-243
Host: Rabbit
Isotype: Ig
Reactivity: Human