Anti-SH3BP2 Rabbit Polyclonal Antibody
Catalog # PRSI13-525
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:SH3-domain binding protein 2
- Antigen symbol:SH3BP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6452
- Antigen synonyms:3BP-2|3BP2|CRPM|SH3BP2|CRBM|RES4-23|FLJ42079
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 392-561 of human SH3BP2 (NP_001116153.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SH3BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat