Anti-SCNN1D Rabbit Polyclonal Antibody
Catalog # PRSI13-524
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:sodium channel, non-voltage-gated 1, delta subunit
- Antigen symbol:SCNN1D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6339
- Antigen synonyms:sodium channel nonvoltage-gated 1 delta|delta polypeptide|ENaCd|delta|DNACH|nonvoltage-gated 1|MGC149711|SCNN1D|SCNED|MGC149710|ENaCdelta|voltage-gated|sodium channel|dNaCh|type I
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 540-680 of human SCNN1D (NP_001123885.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat