Anti-SERPINA3 Rabbit Polyclonal Antibody
Catalog # PRSI13-502
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3
- Antigen symbol:SERPINA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:12
- Antigen synonyms:Cell growth-inhibiting gene 24/25 protein|Alpha-1-antichymotrypsin|Serpin A3|AACT|ACT|Alpha-1-antichymotrypsin His-Pro-less|SERPINA3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 24-423 of human SERPINA3 (NP_001076.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SERPINA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat