Anti-ZSCAN4 Rabbit Polyclonal Antibody
PRSI13-499
- Antibody type:Primary
- Antigen name:zinc finger and SCAN domain containing 4C
- Antigen symbol:ZSCAN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:245109
- Antigen synonyms:XM_142517|Gm397|zinc finger and SCAN domain containing protein 4C|Zscan4d
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of mouse ZSCAN4
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ZSCAN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse