Anti-IARS Rabbit Polyclonal Antibody
Catalog # PRSI13-488
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:isoleucyl-tRNA synthetase
- Antigen symbol:IARS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3376
- Antigen synonyms:isoleucine tRNA ligase 1|IRS|cytoplasmic|GRIDHH1|PRO0785|ILERS|isoleucine--tRNA ligase|IARS|ILRS
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1158-1262 of human IARS (NP_002152.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: IARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat