Anti-BAGE2 Rabbit Polyclonal Antibody
Catalog # PRSI13-472
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:BAGE family member 2
- Antigen symbol:BAGE2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:85319
- Antigen synonyms:cancer/testis antigen family 2|B melanoma antigen family member 2|member 2|cancer/testis antigen 2.2|CT2.2|B melanoma antigen 2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 18-109 of human BAGE2 (NP_872288.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: BAGE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human