Anti-DUSP16 Rabbit Polyclonal Antibody
Catalog # PRSI13-468
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:dual specificity phosphatase 16
- Antigen symbol:DUSP16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:80824
- Antigen synonyms:KIAA1700|MKP7|MAPK phosphatase-7|dual specificity phosphatase 16|MGC129701|MGC129702|DUSP16|MKP-7
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 416-665 of human DUSP16 (NP_085143.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DUSP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat