Anti-C6orf25 Rabbit Polyclonal Antibody
Catalog # PRSI13-467
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:megakaryocyte and platelet inhibitory receptor G6b
- Antigen symbol:C6orf25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:80739
- Antigen synonyms:G6b-B|G6b|THAMY|C6orf25|NG31|megakaryocyte and platelet inhibitory receptor G6b|protein G6b|immunoglobulin receptor
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 18-142 of human C6orf25 (NP_612121.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: C6orf25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat