Anti-RPS6KA4 Rabbit Polyclonal Antibody
Catalog # PRSI13-447
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal protein S6 kinase alpha-4
- Antigen symbol:RPS6KA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:8986
- Antigen synonyms:MSK2|Ribosomal protein kinase B|S6K-alpha-4|RPS6KA4|Nuclear mitogen- and stress-activated protein kinase 2|RSKB|Ribosomal protein S6 kinase alpha-4|90 kDa ribosomal protein S6 kinase 4
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human RPS6KA4 (NP_003933.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RPS6KA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat