Anti-SCN7A Rabbit Polyclonal Antibody
Catalog # PRSI13-423
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:sodium voltage-gated channel alpha subunit 7
- Antigen symbol:SCN7A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6332
- Antigen synonyms:NaG|SCN6A|type VII|sodium channel protein cardiac and skeletal muscle subunit alpha|alpha subunit|Nav2.2|putative voltage-gated sodium channel subunit alpha Nax|voltage-dependent sodium channel alpha subunit|Nav2.1|type VI|alpha polypeptide|sodium channel protein type 7 subunit alpha|voltage-gated|sodium channel|sodium channel protein type VII subunit alpha
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 265-368 of human SCN7A (NP_002967.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SCN7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse