Anti-MAFB Rabbit Polyclonal Antibody
PRSI13-416
- Antibody type:Primary
- Antigen name:v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B
- Antigen symbol:MAFB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:9935
- Antigen synonyms:KRML|MCTO|MAFB|MGC43127
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 184-323 of human MAFB (NP_005452.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MAFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat