Anti-DDR2 Rabbit Polyclonal Antibody
Catalog # PRSI13-402
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:discoidin domain receptor tyrosine kinase 2
- Antigen symbol:DDR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4921
- Antigen synonyms:MIG20a|Hydroxyaryl-protein kinase|NTRKR3|DDR2|Discoidin domain receptor 2|Tyrosylprotein kinase|TYRO10|Tyrosine-protein kinase TYRO10|CD167b antigen
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 290-400 of human DDR2 (NP_001014796.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DDR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat