Anti-SUCLA2 Rabbit Polyclonal Antibody
Codice catalogo PRSI13-392
Nome del fornitore: ProSci Inc.
Specifiche
- Antibody type:Primary
- Antigen name:succinate-CoA ligase ADP-forming beta subunit
- Antigen symbol:SUCLA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:8803
- Antigen synonyms:succinate--CoA ligase [ADP-forming] subunit beta|MTDPS5|succinate-CoA ligase beta subunit|ATP-specific succinyl-CoA synthetase subunit beta|A-BETA|SCS-betaA|mitochondrial|renal carcinoma antigen NY-REN-39|succinyl-CoA ligase [ADP-forming] subunit beta|ATP-specific succinyl-CoA synthetase|mitochondrial succinyl-CoA ligase [ADP-forming] subunit beta|succinyl-CoA synthetase beta-A chain|beta subunit|A-SCS
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human SUCLA2 (NP_003841.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: SUCLA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat