Anti-LRRC59 Rabbit Polyclonal Antibody
Codice catalogo PRSI13-381
Nome del fornitore: ProSci Inc.
Specifiche
- Antibody type:Primary
- Antigen name:leucine rich repeat containing 59
- Antigen symbol:LRRC59
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:55379
- Antigen synonyms:PRO1855|FLJ21675|p34|LRRC59
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-244 of human LRRC59 (NP_060979.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: LRRC59
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat