Anti-HPCAL1 Rabbit Polyclonal Antibody
Catalog # PRSI13-378
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:hippocalcin like 1
- Antigen symbol:HPCAL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:3241
- Antigen synonyms:hippocalcin-like protein 1|VILIP-3|BDR1|HLP2|calcium-binding protein BDR-1|visinin-like protein 3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-193 of human HPCAL1 (NP_002140.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: HPCAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat