Anti-PRKAR1A Rabbit Polyclonal Antibody
Catalog # PRSI13-319
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:protein kinase cAMP-dependent type I regulatory subunit alpha
- Antigen symbol:PRKAR1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5573
- Antigen synonyms:epididymis secretory sperm binding protein|cAMP-dependent protein kinase regulatory subunit RIalpha|alpha|regulatory|CNC1|cAMP-dependent protein kinase type I-alpha regulatory subunit|PKR1|regulatory subunit type I alpha|CNC|protein kinase|PRKAR1|ADOHR|PPNAD1|Carney complex type 1|protein kinase A type 1a regulatory subunit|TSE1|cAMP-dependent|CAR|tissue-specific extinguisher 1|cAMP-dependent protein kinase type I-alpha regulatory chain|type I|ACRDYS1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human PRKAR1A (NP_002725.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PRKAR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human