Anti-GNA15 Rabbit Polyclonal Antibody
Catalog # PRSI18-484
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Guanine nucleotide-binding protein subunit alpha-15
- Antigen symbol:GNA15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2769
- Antigen synonyms:Guanine nucleotide-binding protein subunit alpha-16|GNA15|G alpha-16|G-protein subunit alpha-15|Guanine nucleotide-binding protein subunit alpha-15|GNA16|Epididymis tissue protein Li 17E|G-protein subunit alpha-16|G alpha-15
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-374 of human GNA15 (NP_002059.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GNA15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat