Anti-CREB3L2 Rabbit Polyclonal Antibody
Catalog # PRSI64-051
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cyclic AMP-responsive element-binding protein 3-like protein 2
- Antigen symbol:CREB3L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:490-517 aa
- Form:Liquid
- Gene ID:64764
- Modification:None
- Molecular weight:57 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CREB3L2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 490-517 amino acids from the C-terminal region of human CREB3L2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: CREB3L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 490-517
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse