Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-JUP Rabbit Polyclonal Antibody
Catalog # PRSI63-735
Supplier:  ProSci Inc.
undefined
Anti-JUP Rabbit Polyclonal Antibody
Catalog # PRSI63-735
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Junction plakoglobin
  • Antigen symbol:
    JUP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    636-663 aa
  • Form:
    Liquid
  • Gene ID:
    3728
  • Modification:
    None
  • Molecular weight:
    82 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Shipping temperature:
    4 °C
  • Immunogen:
    This JUP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 636-663 amino acids from the C-terminal region of human JUP.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Pk:
    400 µl

Specifications

About this item

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: JUP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 636-663
Host: Rabbit
Isotype: Ig
Reactivity: Human