Anti-GOT2 Rabbit Polyclonal Antibody
Catalog # PRSI63-724
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Aspartate aminotransferase, mitochondrial
- Antigen symbol:GOT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:33-61 aa
- Form:Liquid
- Gene ID:2806
- Modification:None
- Molecular weight:48 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This GOT2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 33-61 amino acids from the N-terminal region of human GOT2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: GOT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 33-61
Host: Rabbit
Isotype: Ig
Reactivity: Human