Anti-ELAVL1 Rabbit Polyclonal Antibody
Catalog # PRSI63-722
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:ELAV-like protein 1
- Antigen symbol:ELAVL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:1994
- Modification:None
- Molecular weight:36 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ELAVL1 antibody is generated from rabbits immunized with human ELAVL1 recombinant protein.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
For FACS starting dilution is: 1:25. For IHC-P starting dilution is: 1:25. For WB starting dilution is: 1:2000.
Type: Primary
Antigen: ELAVL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse