Anti-ALDH18A1 Rabbit Polyclonal Antibody
PRSI63-710
New Product
- Antibody type:Primary
- Antigen name:Delta-1-pyrroline-5-carboxylate synthase
- Antigen symbol:ALDH18A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:176-205 aa
- Form:Liquid
- Gene ID:5832
- Modification:None
- Molecular weight:87 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,42 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ALDH18A1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 176-205 amino acids from the Central region of human ALDH18A1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: ALDH18A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 176-205
Host: Rabbit
Isotype: Ig
Reactivity: Human