Anti-HNF1A Rabbit Polyclonal Antibody
PRSI63-609
New Product
- Antibody type:Primary
- Antigen name:Hepatocyte nuclear factor 1-alpha
- Antigen symbol:HNF1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:177-205 aa
- Form:Liquid
- Gene ID:6927
- Modification:None
- Molecular weight:67 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This HNF1A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 177-205 amino acids from the Central region of human HNF1A.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:10~50.
Type: Primary
Antigen: HNF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 177-205
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse