Anti-PTPN3 Rabbit Polyclonal Antibody
Catalog # PRSI63-464
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Tyrosine-protein phosphatase non-receptor type 3
- Antigen-Symbol:PTPN3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Mouse
- Western blot:Yes
- Epitop:366-397 aa
- Form:Liquid
- Gen-ID:5774
- Modification:None
- Molecular weight:104 kDa
- Lagertemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Konzentration:2 mg/ml
- Transporttemperatur:4 °C
- Immunogen:This PTPH1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 366-397 amino acids from the Central region of human PTPH1.
- Reinigung:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- VE:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: PTPN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 366-397
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse