Anti-ITPKA Rabbit Polyclonal Antibody
PRSI63-441
- Antibody type:Primary
- Antigen name:Inositol-trisphosphate 3-kinase A
- Antigen symbol:ITPKA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:345-375 aa
- Form:Liquid
- Gene ID:3706
- Modification:None
- Molecular weight:51 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ITPKA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 345-375 amino acids from the C-terminal region of human ITPKA.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: ITPKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 345-375
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse