Anti-NPR3 Rabbit Polyclonal Antibody
PRSI63-401
- Antibody type:Primary
- Antigen name:Atrial natriuretic peptide receptor 3
- Antigen symbol:NPR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:67-97 aa
- Form:Liquid
- Gene ID:4883
- Modification:None
- Molecular weight:60 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Natriuretic Peptide Receptor C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 67-97 amino acids from the N-terminal region of human Natriuretic Peptide Receptor C.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:200 µl
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:200. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: NPR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 67-97
Host: Rabbit
Isotype: Ig
Reactivity: Human