Anti-MAPK13 Rabbit Polyclonal Antibody
Catalog # PRSI62-960
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 13
- Antigen symbol:MAPK13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Epitope:335-365 aa
- Form:Liquid
- Gene ID:5603
- Modification:None
- Molecular weight:42 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This MAPK13 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 335-365 amino acids from the C-terminal region of human MAPK13.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: MAPK13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 335-365
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse, Rat, Monkey