Anti-MSRA Rabbit Polyclonal Antibody
Catalog # PRSI62-820
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Mitochondrial peptide methionine sulfoxide reductase
- Antigen symbol:MSRA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:34-63 aa
- Form:Liquid
- Gene ID:4482
- Modification:None
- Molecular weight:26 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This MSRA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 34-63 amino acids from the N-terminal region of human MSRA.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IF starting dilution is: 1:100. For IHC-P starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: MSRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 34-63
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse