Anti-Nucleocapsid Phosphoprotein Rabbit Recombinant Monoclonal Antibody [clone: 3E8A5]
Catalog # PRSI24-036
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Nucleocapsid phosphoprotein
- Antigen symbol:N
- Clone:3E8A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Form:Lyophilized
- Gene ID:1489678
- Antigen synonyms:2019-nCoV N|2019nCoV N|2019-nCoV Nucleoprotein|2019-nCoV nucleocapsid protein.|SARS-CoV-2 NP|2019 peak N Protein|nucleocapsid protein [Severe acute respiratory syndrome coronavirus 2]|SARS-CoV-2 Nucleocapsid Protein|novel coronavirus Nucleocapsid Protein|novel coronavirus N Protein|2019-nCoV N Protein
- Modification:None
- Storage buffer:Lyophilised from a 0,2 μm filtered solution in PBS, pH 7,4
- Storage temperature:2...8 °C for one month, −20...−80 °C for one year. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:RT
- Immunogen:E. coli derived SARS-COV-2 Nucleoproteins Ser2-Ala419(Gly335Ala) Accession #QHD43423.2
- Purification:Affinity purification 1.0 EU/µg of the rotein by LAL method.
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: N
Clonality: Recombinant Monoclonal
Clone: 3E8A5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus