Anti-CDKN2A Rabbit Polyclonal Antibody
Catalog # PRSI24-023
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:cyclin dependent kinase inhibitor 2A
- Antigen symbol:CDKN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:1029
- Antigen synonyms:CDKN2|MLM|INK4|MTS-1|P16|P16INK4|P19|P16-INK4A|P16INK4A|INK4A|Cyclin-dependent kinase 4 inhibitor A|P19ARF|CDKN2A Antibody: ARF|CDK4I|CMM2|P14|MTS1|TP16
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 20 to the C-terminus of human CDKN2A / p16INK4a
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CDKN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat