Anti-MZT2B Rabbit Polyclonal Antibody
Catalog # PRSI24-012
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:mitotic spindle organizing protein 2B
- Antigen symbol:MZT2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:80097
- Antigen synonyms:family with sequence similarity 128|FAM128B|mitotic-spindle organizing protein 2B|mitotic-spindle organizing protein associated with a ring of gamma-tubulin 2B|MOZART2B|member B
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-158 of human MZT2B (NP_079305.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MZT2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human