Anti-FBLN5 Rabbit Polyclonal Antibody
Catalog # PRSI23-998
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:fibulin 5
- Antigen symbol:FBLN5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10516
- Antigen synonyms:UNQ184/PRO210|ARMD3|EVEC|UP50|FBLN5|developmental arteries and neural crest epidermal growth factor-like|fibulin 5|FLJ90059|DANCE|urine p50 protein
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 199-448 of human FBLN5 (NP_006320.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: FBLN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse