Anti-STRA13 Rabbit Polyclonal Antibody
Catalog # PRSI23-961
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:centromere protein X
- Antigen symbol:STRA13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:201254
- Antigen synonyms:stimulated by retinoic acid gene 13 protein homolog|FANCM associated histone fold protein 2|CENP-X|retinoic acid-inducible gene D9 protein homolog|centromere protein X|FANCM-interacting histone fold protein 2|D9|stimulated by retinoic acid 13 homolog|FAAP1MHF2|STRA13|Fanconi anemia-associated polypeptide of 10 kDa
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-63 of human STRA13 (NP_659435.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: STRA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human