Anti-UBE2NL Rabbit Polyclonal Antibody
PRSI23-545
New Product
- Antibody type:Primary
- Antigen name:ubiquitin conjugating enzyme E2 N like (gene/pseudogene)
- Antigen symbol:UBE2NL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:389898
- Antigen synonyms:epididymis tissue protein Li 174|ubiquitin conjugating enzyme E2N-like|putative ubiquitin-conjugating enzyme E2 N-like|Li174
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human UBE2NL (NP_001013007.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: UBE2NL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse