Anti-VCAM1 Recombinant Antibody [Clone: B-K9 (C106-K9)]
Catalog # PRSI24-772
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:vascular cell adhesion molecule 1
- Antigen symbol:VCAM1
- Clonality:Recombinant
- Clone:B-K9 (C106-K9)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:7412
- Antigen synonyms:V-CAM 1|VCAM1|Vascular cell adhesion protein 1|INCAM-100
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing Balb/c mice against activated human umbilical vein endothelial cells (HUVEC)
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: vascular cell adhesion molecule 1
Clonality: Recombinant
Clone: B-K9 (C106-K9)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human