Anti-CD63 Recombinant Antibody [Clone: NK-1-C3]
PRSI24-761
- Antibody type:Primary
- Antigen name:CD63 molecule
- Antigen symbol:CD63
- Clonality:Recombinant
- Clone:NK-1-C3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:967
- Antigen synonyms:NK-1.C3|Tspan-30|Ocular melanoma-associated antigen|OMA81H|NK1C3|Melanoma-associated antigen ME491|Lysosomal-associated membrane protein 3|Tetraspanin-30|NKI/C3|Granulophysin|LAMP-3|CD63 antigen|NK1/C3
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing BALB/c mice with a plasma membrane preparation of the human melanoma MeWo cells
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: CD63 molecule
Clonality: Recombinant
Clone: NK-1-C3
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human;Mouse