Anti-PECAM1 Recombinant Antibody [Clone: BAG-85D10]
Katalognr. PRSI24-739
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:platelet and endothelial cell adhesion molecule 1
- Antigen symbol:PECAM1
- Clonality:Recombinant
- Clone:BAG-85D10
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:5175
- Antigen synonyms:BAG 85D10|Platelet endothelial cell adhesion molecule|PECAM-1|GPIIA'|EndoCAM|PECAM1|PECA1|JC/70A
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising mice with human peripheral blood lymphocytes from a patient with high numbers of gamma-delta T cells
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: platelet and endothelial cell adhesion molecule 1
Clonality: Recombinant
Clone: BAG-85D10
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human