ContactUsLinkComponent
Tryptone Peptone Glucose Yeast Extract Broth Base
Catalog # HIMEM969-500G
Supplier:  HIMEDIA
CAS Number:  
Tryptone Peptone Glucose Yeast Extract Broth Base
Catalog # HIMEM969-500G
Supplier:  HIMEDIA
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Tryptone peptone glucose yeast extract broth base
Specifications
About this item
This broth base without trypsin is used to test toxicity type of Clostridium botulinum cultures.