Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Bovine trypsin (from pancreas)
Bovine trypsin (from pancreas)
Catalog # CHMCAB1823
Anbieter:
Chemicon
CAS-Nummer:
Expand image
Bovine trypsin (from pancreas)
Catalog # CHMCAB1823
Anbieter:
Chemicon
CAS-Nummer:
Technische Daten
Pk:
25 mg
CAS No.:
9002-07-7
Source:
Pancreas
Species:
Bovine
Storage conditions:
Refrigerator
MDL No.:
MFCD00082094
Enzyme name:
Trypsin
Technische Daten
Über diesen Artikel
Product Details
Product Details